Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-6-cyanopyridine, 97%

Catalog No. AAH2774806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2774806 5 g
AAH2774803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2774806 Supplier Thermo Scientific Chemicals Supplier No. H2774806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33252-29-8 | C6H3ClN2 | 138.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33252-29-8
Molecular Formula C6H3ClN2
Molecular Weight (g/mol) 138.55
MDL Number MFCD00274527
InChI Key PGZHSVWXFKKCNR-UHFFFAOYSA-N
Synonym 2-chloro-6-cyanopyridine, 6-chloropicolinonitrile, 6-chloro-2-cyanopyridine, 2-pyridinecarbonitrile, 6-chloro, 6-chloro-2-pyridinecarbonitrile, 6-chloro-pyridine-2-carbonitrile, 6-chlorpyridin-2-carbonitril, pubchem2163, acmc-1adyc, 2-cyano-6-chloropyridine
PubChem CID 118424
IUPAC Name 6-chloropyridine-2-carbonitrile
SMILES ClC1=CC=CC(=N1)C#N

Specifications

Melting Point 85°C to 88°C
Quantity 5 g
UN Number UN3439
Formula Weight 138.56
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-cyanopyridine
Hazard Category H302+H332-H311-H315-H319-H335
Hazard Statement GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
EINECSNumber 251-429-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.