accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33252-63-0

IUPAC Name: 5-(trifluoromethyl)-1,2-dihydropyridin-2-one
Synonyms: 5-(trifluoromethyl)-1H-pyridin-2-one
Molecular Weight (g/mol): 163.1
Molecular Formula: C6H4F3NO
InChi Key: BYRJSCNPUHYZQE-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CNC(=O)C=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results