accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33263-43-3

NH2SOONCl

CAS RN 33263-43-3

IUPAC Name: 4-chloropyridine-3-sulfonamide
Synonyms: 4-chloropyridine-3-sulfonamide
Molecular Weight (g/mol): 192.62
Molecular Formula: C5H5ClN2O2S
InChi Key: DGIINIBYHCODIH-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CN=CC=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results