Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Chloro-3-pyridinesulfonamide, 98%, Thermo Scientific™

Catalog No. AAL1949306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1949306 5 g
AAL1949303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1949306 Supplier Thermo Scientific Chemicals Supplier No. L1949306

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33263-43-3
Molecular Formula C5H5ClN2O2S
Molecular Weight (g/mol) 192.617
MDL Number MFCD04038136
InChI Key DGIINIBYHCODIH-UHFFFAOYSA-N
Synonym 4-chloro-3-pyridinesulfonamide, 4-chloro-3-pyridinesulphonamide, 3-pyridinesulfonamide, 4-chloro, 4-chlorpyridin-3-sulfonamid, 4-chloropyridine-3-sulphonamide, acmc-20a2oj, 4-chloropyridin-3-sulfonamide, 4-chloro-3-pyridine sulfonamide, 4-chloropyridine-3-sulfonamide, 3-pyridinesulfonamide,4-chloro
PubChem CID 118426
IUPAC Name 4-chloropyridine-3-sulfonamide
SMILES C1=CN=CC(=C1Cl)S(=O)(=O)N

Specifications

Melting Point 150°C to 155°C (decomposition)
Quantity 5 g
Formula Weight 192.62
Percent Purity 98%
Chemical Name or Material 4-Chloro-3-pyridinesulfonamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 251-434-4
TSCA No
Recommended Storage Keep cold
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.