accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFFOFFFFFFFFFFFFF

CAS RN 3330-14-1

IUPAC Name: 1,1,1,2,2,3,3-heptafluoro-3-{[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy}propane
Synonyms: 1,1,1,2,2,3,3-heptafluoro-3-{[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy}propane
Molecular Weight (g/mol): 452.07
Molecular Formula: C8HF17O2
InChi Key: PYSYKOPZHYNYSZ-UHFFFAOYNA-N
SMILES: FC(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)