Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,1,1,2,2,3,3-Heptafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy]propane 90.0+%, TCI America™
SDP

Catalog No. H162425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H162425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H162425G Supplier TCI America Supplier No. H162425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3330-14-1
Molecular Formula C8HF17O2
Molecular Weight (g/mol) 452.067
MDL Number MFCD00054714
InChI Key PYSYKOPZHYNYSZ-UHFFFAOYSA-N
Synonym 2h-perfluoro-5-methyl-3,6-dioxanonane, 1,1,1,2,2,3,3-heptafluoro-3-1,1,1,2,3,3-hexafluoro-3-1,2,2,2-tetrafluoroethoxy propan-2-yl oxy propane, propane, 1-1-difluoro 1,2,2,2-tetrafluoroethoxy methyl-1,2,2,2-tetrafluoroethoxy-1,1,2,2,3,3,3-heptafluoro, cf3cfhocf2cf cf3 ocf2cf2cf3, 2h-perfluoro 5-methyl-3,6-dioxanonane, 1,1,1,2,3,3-hexafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy-3-1,2,2,2-tetrafluoroethoxy propane, 1,1,1,2,4,4,5,7,7,8,8,9,9,9-tetradecafluoro-5-trifluoromethyl-3,6-dioxanonane
PubChem CID 102982
IUPAC Name 1,1,1,2,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)-3-(1,2,2,2-tetrafluoroethoxy)propane
SMILES C(C(F)(F)F)(OC(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)(F)F)F

Specifications

Color Colorless
Boiling Point 105°C
Quantity 25 g
Formula Weight 452.07
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,1,1,2,2,3,3-Heptafluoro-3-[[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy]pr
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.