accessibility menu, dialog, popup

UserName

Viewing profile as user:
FOFFFOFFFOFFFFFFFFFFFFFFFF

CAS RN 3330-16-3

IUPAC Name: 1,1,1,2,2,3,3-heptafluoro-3-[(1,1,1,2,3,3-hexafluoro-3-{[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy}propan-2-yl)oxy]propane
Synonyms: 1,1,1,2,2,3,3-heptafluoro-3-[(1,1,1,2,3,3-hexafluoro-3-{[1,1,1,2,3,3-hexafluoro-3-(1,2,2,2-tetrafluoroethoxy)propan-2-yl]oxy}propan-2-yl)oxy]propane
Molecular Weight (g/mol): 618.09
Molecular Formula: C11HF23O3
InChi Key: QRLOKTBZZPZAMX-UHFFFAOYNA-N
SMILES: FC(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results