CAS RN 33396-29-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Erythromycin A enol ether, ≥97% (HPLC), MilliporeSigma™ Supelco™
Erythromycin A enol ether (EAEN) is an acidic degradation product of erythromycin A (EA) which is grouped under the macrolide antibiotic class of compounds.