accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaNaNH2NONNHOOPOOOOHN

CAS RN 33430-61-4

IUPAC Name: disodium [5-(2-amino-6-oxo-6,9-dihydro-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Synonyms: disodium [5-(2-amino-6-oxo-3H-purin-9-yl)-3-hydroxyoxolan-2-yl]methyl phosphate
Molecular Weight (g/mol): 391.19
Molecular Formula: C10H12N5Na2O7P
InChi Key: CTPAMSRBXKGZCJ-UHFFFAOYNA-L
SMILES: [Na+].[Na+].NC1=NC(=O)C2=C(N1)N(C=N2)C1CC(O)C(COP([O-])([O-])=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results