accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33457-48-6

NNHOHHO

CAS RN 33457-48-6

IUPAC Name: [5-(hydroxymethyl)-1H-imidazol-4-yl]methanol
Synonyms: [5-(hydroxymethyl)-1H-imidazol-4-yl]methanol
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: AGJXDKGTTMVHOU-UHFFFAOYSA-N
SMILES: OCC1=C(CO)N=CN1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results