accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 334869-71-5

IUPAC Name: neodymium(3+) triacetate
Synonyms: neodymium triacetate
Molecular Weight (g/mol): 321.37
Molecular Formula: C6H9NdO6
InChi Key: KETUDCKOKOGBJB-UHFFFAOYSA-K
SMILES: [Nd+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results