Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Trimethylphosphine Solution

Catalog No. 324116100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-411-6100ML 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 32-411-6100ML Supplier Sigma Aldrich Supplier No. 324116100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reagent used in Mitsunobu reaction; transformation of azides into carbamates, aziridines from azidoalcohols; iminophosphoranes and aza-Wittig reaction.

Specifications

CAS 334869-71-5
Molecular Formula C6H9NdO6 -+ xH2O
Linear Formula (CH3CO2)3Nd -+ xH2O
MDL Number MFCD00150120
Quantity 50 g
Synonym PMe3; Trimethylphosphorus
Molecular Weight (g/mol) 321.37 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.