accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 334949-59-6

IUPAC Name: 3-[({3-[(dimethylamino)methyl]phenyl}imino)(phenyl)methyl]-2-hydroxy-1H-indole-6-carboxamide
Synonyms: 3-[({3-[(dimethylamino)methyl]phenyl}imino)(phenyl)methyl]-2-hydroxy-1H-indole-6-carboxamide
Molecular Weight (g/mol): 412.49
Molecular Formula: C25H24N4O2
InChi Key: WGPXKFOFEXJMBD-UHFFFAOYSA-N
SMILES: CN(C)CC1=CC(=CC=C1)N=C(C1=C(O)NC2=CC(=CC=C12)C(N)=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results