accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSOONH2

CAS RN 334981-10-1

IUPAC Name: 4-[(pyrrolidine-1-sulfonyl)methyl]aniline
Synonyms: 4-[(pyrrolidine-1-sulfonyl)methyl]aniline
Molecular Weight (g/mol): 240.32
Molecular Formula: C11H16N2O2S
InChi Key: VNSKHALYBQZMFW-UHFFFAOYSA-N
SMILES: NC1=CC=C(CS(=O)(=O)N2CCCC2)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results