accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 335-48-8

FFBrFFFFBrFF

CAS RN 335-48-8

IUPAC Name: 1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
Synonyms: 1,4-dibromooctafluorobutane
Molecular Weight (g/mol): 359.84
Molecular Formula: C4Br2F8
InChi Key: RWWUGYJWSVESJC-UHFFFAOYSA-N
SMILES: FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)