Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dibromooctafluorobutane 98.0+%, TCI America™
SDP

Catalog No. D35735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D35735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D35735G Supplier TCI America Supplier No. D35735G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 335-48-8
Molecular Formula C4Br2F8
Molecular Weight (g/mol) 359.839
MDL Number MFCD00153112
InChI Key RWWUGYJWSVESJC-UHFFFAOYSA-N
Synonym 1,4-dibromooctafluorobutane, 1,4-dibromoperfluorobutane, butane, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, acmc-209i1o, 1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro-butane, butane,1,4-dibromo-1,1,2,2,3,3,4,4-octafluoro, 1,4-bis bromanyl-1,1,2,2,3,3,4,4-octakis fluoranyl butane
PubChem CID 2736783
IUPAC Name 1,4-dibromo-1,1,2,2,3,3,4,4-octafluorobutane
SMILES C(C(C(F)(F)Br)(F)F)(C(F)(F)Br)(F)F

Specifications

Color Yellow
Boiling Point 104°C
Quantity 5 g
Formula Weight 359.84
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,4-Dibromooctafluorobutane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.