accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 335-58-0

IUPAC Name: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
Synonyms: 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
Molecular Weight (g/mol): 495.96
Molecular Formula: C7F15I
InChi Key: AHUMDLIBMIYQMU-UHFFFAOYSA-N
SMILES: FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results