Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pentadecafluoroheptyl Iodide (stabilized with Na2S2O3) 98.0+%, TCI America™
SDP

Catalog No. P18395G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18395G 5 g
P183925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. P18395G Supplier TCI America Supplier No. P18395G

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 335-58-0
Molecular Formula C7F15I
Molecular Weight (g/mol) 495.958
MDL Number MFCD00013712
InChI Key AHUMDLIBMIYQMU-UHFFFAOYSA-N
Synonym Perfluoroheptyl Iodide
PubChem CID 67633
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7-pentadecafluoro-7-iodoheptane
SMILES C(C(C(C(F)(F)F)(F)F)(F)F)(C(C(C(F)(F)I)(F)F)(F)F)(F)F

Specifications

Melting Point -8°C
Color Yellow
Boiling Point 138°C
Quantity 5 g
Formula Weight 495.96
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pentadecafluoroheptyl Iodide (stabilized with Na2S2O3)
EINECSNumber (2)-0090
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.