accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 335033-68-6

IUPAC Name: methyl({4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methyl)amine hydrochloride
Synonyms: methyl({4-methylthieno[3,2-b]pyrrol-5-yl}methyl)amine hydrochloride
Molecular Weight (g/mol): 216.73
Molecular Formula: C9H13ClN2S
InChi Key: JUWOUDJRMHOUBO-UHFFFAOYSA-N
SMILES: Cl.CNCC1=CC2=C(C=CS2)N1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results