Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl-N-[(4-methyl-4H-thieno[3,2-b]pyrrol-5-yl)methyl]amine hydrochloride, 95%, Thermo Scientific™

Catalog No. CC40996DA
Change view
Click to view available options
Amber Glass Bottle; 1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
CC40996DA Amber Glass Bottle; 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC40996DA Supplier Thermo Scientific Chemicals Supplier No. CC40996DA

missing translation for 'validMainframeMsg'

Specifications

CAS 335033-68-6
CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C9H13ClN2S
Quantity 1 g
MDL Number MFCD07772840
Synonym n-methyl-n-4-methyl-4h-thieno 3,2-b pyrrol-5-yl methyl amine hydrochloride, methyl 4-methylthieno 3,2-b pyrrol-5-yl methyl amine hydrochloride, methyl 4-methylthiopheno 2,3-d pyrrol-5-yl methyl amine, chloride, n-methyl-n-4-methyl-4h-theno 3,2-b pyrrol-5-yl methyl amne hydrochlorde, n-methyl-1-4-methyl-4h-thieno 3,2-b pyrrol-5-yl methanamine-hydrogen chloride 1/1
InChI Key JUWOUDJRMHOUBO-UHFFFAOYSA-N
SMILES Cl.CNCC1=CC2=C(C=CS2)N1C
IUPAC Name methyl({4-methyl-4H-thieno[3,2-b]pyrrol-5-yl}methyl)amine hydrochloride
Molecular Weight (g/mol) 216.73
PubChem CID 18525750
Formula Weight 216.73
Percent Purity 95%
Chemical Name or Material N-Methyl-N-[(4-methyl-4H-thieno[3, 2-b]pyrrol-5-yl)methyl]amine hydrochloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful in contact with skin.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.