accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 335166-00-2

NHNNBrBrFHClHCl

CAS RN 335166-00-2

IUPAC Name: 3,6-dibromo-9-[2-fluoro-3-(piperazin-1-yl)propyl]-9H-carbazole dihydrochloride
Synonyms: 3,6-dibromo-9-[2-fluoro-3-(piperazin-1-yl)propyl]carbazole dihydrochloride
Molecular Weight (g/mol): 542.11
Molecular Formula: C19H22Br2Cl2FN3
InChi Key: JWCVUOLQOHIRML-UHFFFAOYNA-N
SMILES: Cl.Cl.FC(CN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2)CN1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results