CAS RN 33528-13-1
IUPAC Name:
N-benzyl-4-methyl-N'-oxobenzene-1-sulfonohydrazide
Synonyms:
N-benzyl-4-methyl-N'-oxobenzenesulfonohydrazide
Molecular Weight (g/mol):
290.34
Molecular Formula:
C14H14N2O3S
InChi Key:
ZXELPGWSCCGNDS-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C=C1)S(=O)(=O)N(CC1=CC=CC=C1)N=O
1
–
1
of
1
results
1
N-Benzyl-N-nitroso-p-toluenesulfonamide 98.0+%, TCI America™
CAS: 33528-13-1 Molecular Formula: C14H14N2O3S Molecular Weight (g/mol): 290.337 MDL Number: MFCD00216617 InChI Key: ZXELPGWSCCGNDS-UHFFFAOYSA-N PubChem CID: 4118076 IUPAC Name: N-benzyl-4-methyl-N-nitrosobenzenesulfonamide SMILES: CC1=CC=C(C=C1)S(=O)(=O)N(CC2=CC=CC=C2)N=O