Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzyl-N-nitroso-p-toluenesulfonamide 98.0+%, TCI America™
SDP

Catalog No. B30515G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30515G 5 g
B305125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30515G Supplier TCI America Supplier No. B30515G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 33528-13-1
Molecular Formula C14H14N2O3S
Molecular Weight (g/mol) 290.337
MDL Number MFCD00216617
InChI Key ZXELPGWSCCGNDS-UHFFFAOYSA-N
PubChem CID 4118076
IUPAC Name N-benzyl-4-methyl-N-nitrosobenzenesulfonamide
SMILES CC1=CC=C(C=C1)S(=O)(=O)N(CC2=CC=CC=C2)N=O

Specifications

Melting Point 85°C
Color Yellow
Quantity 5 g
Formula Weight 290.34
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzyl-N-nitroso-p-toluenesulfonamide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.