accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOFFFFFFFFOHO

CAS RN 336-08-3

IUPAC Name: octafluorohexanedioic acid
Synonyms: perfluoroadipic acid
Molecular Weight (g/mol): 290.07
Molecular Formula: C6H2F8O4
InChi Key: AXRSOGFYDSXLQX-UHFFFAOYSA-N
SMILES: OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results