Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octafluoroadipic Acid 98.0+%, TCI America™
SDP

Catalog No. O02605G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02605G 5 g
O026025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02605G Supplier TCI America Supplier No. O02605G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 336-08-3
Molecular Formula C6H2F8O4
Molecular Weight (g/mol) 290.07
MDL Number MFCD00066405
InChI Key AXRSOGFYDSXLQX-UHFFFAOYSA-N
Synonym Octafluorohexanedioic Acid, Perfluoroadipic Acid, Perfluorohexanedioic Acid
PubChem CID 67640
IUPAC Name octafluorohexanedioic acid
SMILES OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(O)=O

Specifications

Melting Point 134°C
Color White-Yellow
Quantity 5 g
Formula Weight 290.07
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Octafluoroadipic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.