accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3360-16-5

IUPAC Name: (3-aminopropyl)(propan-2-yl)amine
Synonyms: (3-aminopropyl)(isopropyl)amine
Molecular Weight (g/mol): 116.21
Molecular Formula: C6H16N2
InChi Key: KFDIDIIKNMZLRZ-UHFFFAOYSA-N
SMILES: CC(C)NCCCN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results