Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Isopropyl-1,3-diaminopropane 98.0+%, TCI America™
SDP

Catalog No. I042025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I042025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I042025ML Supplier TCI America Supplier No. I042025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3360-16-5
Molecular Formula C6H16N2
Molecular Weight (g/mol) 116.208
MDL Number MFCD00008208
InChI Key KFDIDIIKNMZLRZ-UHFFFAOYSA-N
Synonym 3-(Isopropylamino)propylamine, N-Isopropyl-1,3-propanediamine
PubChem CID 76888
IUPAC Name N'-propan-2-ylpropane-1,3-diamine
SMILES CC(C)NCCCN

Specifications

Color Yellow
Boiling Point 162°C
Quantity 25 mL
UN Number 2734
Formula Weight 116.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Isopropyl-1,3-diaminopropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.