accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 336879-53-9

IUPAC Name: magnesium(2+) bis((2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate) hydrate
Synonyms: magnesium(2+) bis(D-gluconate) hydrate
Molecular Weight (g/mol): 432.61
Molecular Formula: C12H24MgO15
InChi Key: IAKLPCRFBAZVRW-XRDLMGPZSA-L
SMILES: O.[Mg++].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O.OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results