accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 33733-73-2

IUPAC Name: 1-bromo-3-(methylsulfanyl)benzene
Synonyms: 1-bromo-3-(methylsulfanyl)benzene
Molecular Weight (g/mol): 203.1
Molecular Formula: C7H7BrS
InChi Key: NKYFJZAKUPSUSH-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(Br)=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)