accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSSOH

CAS RN 337508-70-0

IUPAC Name: [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
Synonyms: [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
Molecular Weight (g/mol): 211.3
Molecular Formula: C9H9NOS2
InChi Key: CQCKNGVVCKWDLF-UHFFFAOYSA-N
SMILES: CC1=NC(=CS1)C1=CC=C(CO)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results