Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[5-(2-Methyl-1,3-thiazol-4-yl)-2-thienyl]methanol 95%, Thermo Scientific™

Catalog No. CC18209EB
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC18209EB 25 g
CC18209DA 1 g
CC18209DE 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC18209EB Supplier Thermo Scientific Chemicals Supplier No. CC18209EB
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 337508-70-0
Molecular Formula C9H9NOS2
Molecular Weight (g/mol) 211.297
InChI Key CQCKNGVVCKWDLF-UHFFFAOYSA-N
Synonym 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methanol, 5-2-methyl-1,3-thiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-4-thiazolyl thiophene-2-methanol, 5-2-methylthiazol-4-yl thiophen-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl thien-2-yl methanol, 5-2-methyl-1,3-thiazol-4-yl-2-thienyl methan-1-ol
PubChem CID 2776470
IUPAC Name [5-(2-methyl-1,3-thiazol-4-yl)thiophen-2-yl]methanol
SMILES CC1=NC(=CS1)C2=CC=C(S2)CO

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 211.31
Percent Purity 95%
Chemical Name or Material [5-(2-Methyl-1, 3-thiazol-4-yl)-2-thienyl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.