accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClOClCl

CAS RN 3380-34-5

IUPAC Name: 5-chloro-2-(2,4-dichlorophenoxy)phenol
Synonyms: triclosan triclosanum manusept irgasan DP300 epa pesticide chemical code 054901 triclosan reckitt brand lexol 300 trans canaderm brand of triclosan irgasan dp 300 cliniclean
Molecular Weight (g/mol): 289.54
Molecular Formula: C12H7Cl3O2
InChi Key: XEFQLINVKFYRCS-UHFFFAOYSA-N
SMILES: OC1=CC(Cl)=CC=C1OC1=CC=C(Cl)C=C1Cl
Percent Purity 
  • (3)
  • (1)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (6)
  • (1)
Boiling Point 
  • (2)
  • (3)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (6)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)