Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triclosan, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog No. 18607559 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-559 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-559 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T158925KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T1589, 3380-34-5, C12H7Cl3O2

Spectrum™ Chemical Triclosan, USP is used as an antibacterial and antifungal agent, as well as a selective agent in molecular cloning. The USP grade indicates it is graded suitable for personal care, cosmetic and pharmaceutical applications. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 3380-34-5
Molecular Formula C12H7Cl3O2
Molecular Weight (g/mol) 289.54
InChI Key XEFQLINVKFYRCS-UHFFFAOYSA-N
IUPAC Name 5-chloro-2-(2,4-dichlorophenoxy)phenol
SMILES OC1=CC(Cl)=CC=C1OC1=CC=C(Cl)C=C1Cl

Specifications

CAS Max % 1
Boiling Point 280 to 290°C
Packaging Fiber Drum
Quantity 25 kg
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.