accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3381-87-1

IUPAC Name: [2-(benzyloxy)phenyl]methanol
Synonyms: [2-(benzyloxy)phenyl]methanol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: XRZMRABYCSOXIZ-UHFFFAOYSA-N
SMILES: OCC1=CC=CC=C1OCC1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results