Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Benzyloxybenzyl Alcohol 97.0+%, TCI America™
SDP

Catalog No. B29435G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B29435G 5 g
B294325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B29435G Supplier TCI America Supplier No. B29435G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3381-87-1
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.264
InChI Key XRZMRABYCSOXIZ-UHFFFAOYSA-N
PubChem CID 18811
IUPAC Name (2-phenylmethoxyphenyl)methanol
SMILES C1=CC=C(C=C1)COC2=CC=CC=C2CO

Specifications

Color White-Yellow
Boiling Point 155°C
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥97.0% (GC)
Physical Form Crystals
Chemical Name or Material 2-Benzyloxybenzyl Alcohol
RTECSNumber DN8582000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.