accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 33838-52-7

IUPAC Name: heptane
Synonyms: heptane N-heptane eptani skellysolve C dipropyl methane gettysolve-C hsdb 90
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: IMNFDUFMRHMDMM-UHFFFAOYSA-N
SMILES: CCCCCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results