Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Heptane-d{16}, 98% (Isotopic)

Catalog No. AA4228903
Change view
Click to view available options
Quantity:
1 g
Packaging:
Prescored Ampule
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA4228903 1 g Prescored Ampule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA4228903 Supplier Thermo Scientific Chemicals Supplier No. 04228903
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 33838-52-7 | C7H16 | 100.21 g/mol

n-Heptane-d{16} is used as a test fuel component in anti-knock test engines, a 100% heptane fuel is the zero point of the octane rating scale. Heptane is commercially available as mixed isomers for use in paints and coatings, as the rubber cement solvent, n-heptane for research and development and pharmaceutical manufacturing and as a minor component of gasoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33838-52-7
Molecular Formula C7H16
Molecular Weight (g/mol) 100.21
MDL Number MFCD00062744
InChI Key IMNFDUFMRHMDMM-UHFFFAOYSA-N
Synonym heptane-d16, n-heptane-d 16, 2h16 heptane, hexadecadeuterio-heptane, 2 h?? heptane, heptane-d16, 99 atom % d
PubChem CID 524600
IUPAC Name 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,7-hexadecadeuterioheptane
SMILES CCCCCCC

Specifications

Physical Form Liquid
Packaging Prescored Ampule
Quantity 1 g
Density 0.794 g/mL
Sensitivity Hygroscopic
Refractive Index 1.3877
Flash Point −4°C (24°F)
Melting Point -91°C to -89°C
Boiling Point 97°C to 99°C
UN Number UN1206
Beilstein 1730763
Solubility Information Difficult to mix.
Formula Weight 116.33
Percent Purity 98%
Chemical Name or Material n-Heptane-d{16}, Isotopic
Show More Show Less
Hazard Category H224-H304-H315-H319-H336
Hazard Statement GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
EINECSNumber 205-563-8
RTECSNumber MI7700000
TSCA No
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.