accessibility menu, dialog, popup

UserName

Viewing profile as user:

Isocyanide dichlorides

Organic compounds that contain a nitrogen atom bonded to a carbon atom that is subsequently bonded to two chlorine atoms in the following structure: R-N-C-Cl2, where R is any carbon chain or group.
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (2)
Physical Form 
  • (5)
Showing products from the Isocyanide dichlorides category. View All Search Results

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (5)