accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3386-18-3

IUPAC Name: 3,6,9,12,15,18,21,24-octaoxahexacosane-1,26-diol
Synonyms: nonaethylene glycol
Molecular Weight (g/mol): 414.49
Molecular Formula: C18H38O10
InChi Key: YZUUTMGDONTGTN-UHFFFAOYSA-N
SMILES: OCCOCCOCCOCCOCCOCCOCCOCCOCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results