Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nonaethylene Glycol 95.0+%, TCI America™
SDP

Catalog No. N06981G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N06981G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N06981G Supplier TCI America Supplier No. N06981G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3386-18-3
Molecular Formula C18H38O10
Molecular Weight (g/mol) 414.492
MDL Number MFCD00698692
InChI Key YZUUTMGDONTGTN-UHFFFAOYSA-N
PubChem CID 4867
ChEBI CHEBI:39784
IUPAC Name 2-[2-[2-[2-[2-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
SMILES C(COCCOCCOCCOCCOCCOCCOCCOCCO)O

Specifications

Color White-Yellow
Quantity 1 g
Physical Form Wax at 20°C
Formula Weight 414.49
Percent Purity ≥95.0% (GC)
Chemical Name or Material Nonaethylene Glycol
EINECSNumber (2)-0441
RTECSNumber RA5400000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.