accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3PH3CCH3H3COOPCH3H3CCH3H3CCH3H3C

CAS RN 338800-20-7

IUPAC Name: 3-[(di-tert-butylphosphanyl)oxy]phenyl di-tert-butylphosphinite
Synonyms: 3-[(di-tert-butylphosphanyl)oxy]phenyl di-tert-butylphosphinite
Molecular Weight (g/mol): 398.51
Molecular Formula: C22H40O2P2
InChi Key: WSKMTURBCLMNEL-UHFFFAOYSA-N
SMILES: CC(C)(C)P(OC1=CC(OP(C(C)(C)C)C(C)(C)C)=CC=C1)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results