Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Bis[(di-tert-butylphosphino)oxy]benzene 94.0+%, TCI America™
SDP

Catalog No. B45951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B45951G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B45951G Supplier TCI America Supplier No. B45951G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 338800-20-7
Molecular Formula C22H40O2P2
Molecular Weight (g/mol) 398.51
MDL Number MFCD28975098
InChI Key WSKMTURBCLMNEL-UHFFFAOYSA-N
PubChem CID 12992261
IUPAC Name 3-[(di-tert-butylphosphanyl)oxy]phenyl di-tert-butylphosphinite
SMILES CC(C)(C)P(OC1=CC(OP(C(C)(C)C)C(C)(C)C)=CC=C1)C(C)(C)C

Specifications

Melting Point 50°C
Color White-Yellow
Quantity 1 g
Formula Weight 398.51
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3-Bis[(di-tert-butylphosphino)oxy]benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.