CAS RN 3391-99-9
CAS RN 3391-99-9
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate
Molecular Weight (g/mol):
362.38
Molecular Formula:
C18H22N2O6
InChi Key:
XQPVVBMUTQVELT-UHFFFAOYNA-N
SMILES:
CCC(C)C(NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Advanced Chem Tech Z-Ile-OSu, Advanced ChemTech
CAS: 3391-99-9 Molecular Formula: C18H22N2O6 Molecular Weight (g/mol): 362.38 MDL Number: MFCD00079002 InChI Key: XQPVVBMUTQVELT-UHFFFAOYNA-N IUPAC Name: 2,5-dioxopyrrolidin-1-yl 2-{[(benzyloxy)carbonyl]amino}-3-methylpentanoate SMILES: CCC(C)C(NC(=O)OCC1=CC=CC=C1)C(=O)ON1C(=O)CCC1=O