CAS RN 3392-07-2
IUPAC Name:
2,5-dioxopyrrolidin-1-yl 2-{[(tert-butoxy)carbonyl]amino}acetate
Synonyms:
2,5-dioxopyrrolidin-1-yl 2-[(tert-butoxycarbonyl)amino]acetate
Molecular Weight (g/mol):
272.26
Molecular Formula:
C11H16N2O6
InChi Key:
LJCWRJYVPJJTMB-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)NCC(=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Promotions Available
N-Boc-glycine N-succinimidyl ester, 98%, Thermo Scientific Chemicals
N-Boc-glycine N-succinimidyl ester, 98%, Thermo Scientific Chemicals
CAS: 3392-07-2