Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Boc-glycine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

Catalog No. AAH6252806
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6252806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6252806 Supplier Thermo Scientific Chemicals Supplier No. H6252806
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 3392-07-2 | Molecular Formula: C11H16N2O6 | Molecular Weight: 272.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3392-07-2

Specifications

Melting Point 165°C to 167°C
Molecular Formula C15H24N2O6
MDL Number MFCD00022585 MFCD00022585
Quantity 5 g
Beilstein 1549242
Sensitivity Hygroscopic
Synonym boc-ile-osu, boc-l-isoleucine hydroxysuccinimide ester, boc-l-isoleucine n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoate, boc-lle-osu, boc-ile-osu hplc, n-alpha-t-butyloxycarbonyl-l-isoleucine succinimidyl ester, 1-n-1,1-dimethylethoxy carbonyl-l-isoleucyl oxy-2,5-pyrrolidinedione, 2,5-dioxopyrrolidin-1-yl 2s,3s-3-methyl-2-2-methylpropan-2-yl oxycarbonylamino pentanoate
InChI Key FATJLEZSGFVHQA-CABZTGNLSA-N
SMILES CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate
Molecular Weight (g/mol) 328.37
PubChem CID 131632401
Formula Weight 272.25
Percent Purity 98%
Chemical Name or Material N-Boc-glycine N-succinimidyl ester
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 222-230-2
TSCA No
Recommended Storage Store at -20°C; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.