CAS RN 3392-08-3
CAS RN 3392-08-3
IUPAC Name:
2,5-dioxopyrrolidin-1-yl (2S,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoate
Synonyms:
2,5-dioxopyrrolidin-1-yl (2S,3S)-2-[(tert-butoxycarbonyl)amino]-3-methylpentanoate
Molecular Weight (g/mol):
328.37
Molecular Formula:
C15H24N2O6
InChi Key:
FATJLEZSGFVHQA-CABZTGNLSA-N
SMILES:
CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
1
–
1
of
1
results
1
Advanced Chem Tech Boc-Ile-OSu, Advanced ChemTech
CAS: 3392-08-3 Molecular Formula: C15H24N2O6 Molecular Weight (g/mol): 328.37 MDL Number: MFCD00022585 MFCD00022585 InChI Key: FATJLEZSGFVHQA-CABZTGNLSA-N IUPAC Name: 2,5-dioxopyrrolidin-1-yl (2S,3S)-2-{[(tert-butoxy)carbonyl]amino}-3-methylpentanoate SMILES: CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O