accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO

CAS RN 3393-45-1

IUPAC Name: 3,6-dihydro-2H-pyran-2-one
Synonyms: 3,6-dihydropyran-2-one
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: VREFDQNWPNDZEX-UHFFFAOYSA-N
SMILES: O=C1CC=CCO1
Boiling Point 
  • (2)
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)