Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5,6-Dihydro-2H-pyran-2-one

Catalog No. 2621021G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-210-21G 1 g
26-210-210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 26-210-21G Supplier Sigma Aldrich Supplier No. 2621021G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Enantioselective conjugate addition of grignard reagents to 5,6-dihydro-2h-pyran-2-one catalyzed by a chiral phosphine-copper iodide catalyst has been reported.5,6-dihydro-2h-pyran-2-one has been used in the preparation of:
  • (1ar,5ar,5bs,6s,7s)-6,7-di-tert-butoxy-5-oxo-pyrrolidino[1,2-b]isoxazolidino[4,5-c]tetrahydropyran
  • 4-(phenyl)tetrahydro-2h-pyran-2-one

Specifications

CAS 3393-45-1
Density 1.139 g/mL (at 25°C (literature))
Boiling Point 103°C (10 mmHg)
Molecular Formula C5H6O2
Refractive Index n20/D 1.483 (literature)
Linear Formula C5H6O2
MDL Number MFCD00010439
Quantity 1 g
Molecular Weight (g/mol) 98.1
Percent Purity 90%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.