accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOHNO

CAS RN 3393-96-2

IUPAC Name: N-(4-nitrophenyl)benzamide
Synonyms: N-(4-nitrophenyl)benzamide
Molecular Weight (g/mol): 242.23
Molecular Formula: C13H10N2O3
InChi Key: GMGQGZYFQSCZCW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(NC(=O)C2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results