Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Nitrobenzanilide 98.0+%, TCI America™
SDP

Catalog No. N039625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N039625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N039625G Supplier TCI America Supplier No. N039625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3393-96-2
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00024619
InChI Key GMGQGZYFQSCZCW-UHFFFAOYSA-N
Synonym N-(4-Nitrophenyl)benzamide
PubChem CID 76930
IUPAC Name N-(4-nitrophenyl)benzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 199°C
Color Yellow
Quantity 25 g
Formula Weight 242.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitrobenzanilide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.